ENA238712782 Enamine

[(3,4-dichlorophenyl)methyl](3-methylbutyl)amine

  • MDL number MFCD09688600
  • Cas Number : 756475-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51211

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51211
Availability details for
Quantity
  • Availability
  • Add to Cart