ENA238712784 Enamine

{3-[(dimethylamino)methyl]phenyl}methanol

  • MDL number MFCD09693970
  • Cas Number : 69383-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51214
Availability details for
Quantity
  • Availability
  • Add to Cart