ENA238712793 Enamine

4-[(azepan-1-yl)methyl]aniline dihydrochloride

  • MDL number MFCD12912831
  • Cas Number : 1193387-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50308
Availability details for
Quantity
  • Availability
  • Add to Cart