ENA238712795 Enamine

N-cyclopropylpiperidine-4-carboxamide

  • MDL number MFCD08753840
  • Cas Number : 937683-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51248
Availability details for
Quantity
  • Availability
  • Add to Cart