ENA238712798 Enamine

3-methylpiperidine-1-carbothioamide

  • MDL number MFCD09736446
  • Cas Number : 954271-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51252
Availability details for
Quantity
  • Availability
  • Add to Cart