ENA238712802 Enamine

(2-methoxyethyl)(1-methoxypropan-2-yl)amine

  • MDL number MFCD11151272
  • Cas Number : 1021042-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51256
Availability details for
Quantity
  • Availability
  • Add to Cart