ENA238712819 Enamine

3-(1-methyl-1H-1,2,3,4-tetrazol-5-yl)benzene-1-sulfonamide

  • MDL number MFCD11650717
  • Cas Number : 1099660-81-7
  • Molecular Weight : 239.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51303
Availability details for
Quantity
  • Availability
  • Add to Cart