ENA238712827 Enamine

3-amino-N-(butan-2-yl)propanamide

  • MDL number MFCD09731731
  • Cas Number : 953890-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51313
Availability details for
Quantity
  • Availability
  • Add to Cart