ENA238712833 Enamine

5-methylbicyclo[3.3.1]nonane-1-carboxylic acid

  • MDL number MFCD00497987
  • Cas Number : 91965-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51320
Availability details for
Quantity
  • Availability
  • Add to Cart