ENA238712845 Enamine

3-aminopropane-1-sulfonamide hydrochloride

  • MDL number MFCD12912951
  • Cas Number : 104458-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51335

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart