ENA238712847 Enamine

5-cyclopropyl-2H-1,2,3,4-tetrazole

  • MDL number MFCD00221169
  • Cas Number : 27943-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51338

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51338
Availability details for
Quantity
  • Availability
  • Add to Cart