ENA238712858 Enamine

2-(2,6-dichlorophenyl)-3-oxobutanenitrile

  • MDL number MFCD11208554
  • Cas Number : 186196-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50316
Availability details for
Quantity
  • Availability
  • Add to Cart