ENA238712863 Enamine

N-[3-(aminomethyl)phenyl]-2-(1H-pyrazol-1-yl)acetamide

  • MDL number MFCD09734119
  • Cas Number : 1052562-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51377

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51377
Availability details for
Quantity
  • Availability
  • Add to Cart