ENA238712870 Enamine

2-cyclopentylpyrrolidine

  • MDL number MFCD02663460
  • Cas Number : 383127-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51385

Safety Information

signalword Danger
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-51385
pictograms GHS05,GHS07
hstatements H227,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart