ENA238712876 Enamine

3-(pyrrolidin-1-ylmethyl)benzonitrile

  • MDL number MFCD09950280
  • Cas Number : 321198-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51391

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51391
Availability details for
Quantity
  • Availability
  • Add to Cart