ENA238712888 Enamine

1-(2-methylpropanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09047850
  • Cas Number : 23500-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51419
Availability details for
Quantity
  • Availability
  • Add to Cart