ENA238712892 Enamine

5-cyclopentyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD11156010
  • Cas Number : 441024-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51423
Availability details for
Quantity
  • Availability
  • Add to Cart