ENA238712893 Enamine

2-(ethylamino)acetic acid hydrochloride

  • MDL number MFCD12912960
  • Cas Number : 542-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51424

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51424
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart