ENA238712906 Enamine

{3-[(1H-pyrazol-1-yl)methyl]phenyl}methanamine

  • MDL number MFCD08572164
  • Cas Number : 562803-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51438
Availability details for
Quantity
  • Availability
  • Add to Cart