ENA238712908 Enamine

1-(2,4-dimethyl-1,3-thiazol-5-yl)ethan-1-amine

  • MDL number MFCD12912964
  • Cas Number : 120276-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51440
Availability details for
Quantity
  • Availability
  • Add to Cart