ENA238712925 Enamine

5-(2,3-dihydro-1-benzofuran-2-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09814066
  • Cas Number : 1016504-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51465
Availability details for
Quantity
  • Availability
  • Add to Cart