ENA238712927 Enamine

(4-{[methyl(propan-2-yl)amino]methyl}phenyl)methanamine dihydrochloride

  • MDL number MFCD12912969
  • Cas Number : 1193388-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51467
Availability details for
Quantity
  • Availability
  • Add to Cart