ENA238712928 Enamine

4,5-dimethoxybenzene-1,2-diamine

  • MDL number MFCD00462577
  • Cas Number : 27841-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-132
Purity 95
Storage temp. -20
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51468
Availability details for
Quantity
  • Availability
  • Add to Cart