ENA238712929 Enamine

4-(5-chlorothiophen-2-yl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD11641427
  • Cas Number : 1096815-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51469

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51469
Availability details for
Quantity
  • Availability
  • Add to Cart