ENA238712930 Enamine

N1-(1,3-benzothiazol-2-yl)benzene-1,3-diamine dihydrochloride

  • MDL number MFCD12912970
  • Cas Number : 1181522-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51470
Availability details for
Quantity
  • Availability
  • Add to Cart