ENA238712942 Enamine

4-{[(4-tert-butylphenyl)methyl]amino}butanoic acid hydrochloride

  • MDL number MFCD12912973
  • Cas Number : 1193388-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51489
Availability details for
Quantity
  • Availability
  • Add to Cart