ENA238712953 Enamine

1-methyl-2-(piperidin-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD05190108
  • Cas Number : 526183-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51505
Availability details for
Quantity
  • Availability
  • Add to Cart