ENA238712955 Enamine

1-cyclopentylpiperidin-4-one

  • MDL number MFCD07374376
  • Cas Number : 343787-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51507
Availability details for
Quantity
  • Availability
  • Add to Cart