ENA238712960 Enamine

1-(3-aminophenyl)-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD09813556
  • Cas Number : 208927-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50331
Availability details for
Quantity
  • Availability
  • Add to Cart