ENA238712968 Enamine

(2,3-dihydro-1H-inden-5-yl)thiourea

  • MDL number MFCD01935531
  • Cas Number : 117174-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51530
Availability details for
Quantity
  • Availability
  • Add to Cart