ENA238712969 Enamine

4-bromo-N'-hydroxythiophene-2-carboximidamide

  • MDL number MFCD11179480
  • Cas Number : 1094456-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51531
Availability details for
Quantity
  • Availability
  • Add to Cart