ENA238712970 Enamine

1-(4-chlorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol

  • MDL number MFCD00690346
  • Cas Number : 62881-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50334
Availability details for
Quantity
  • Availability
  • Add to Cart