ENA238712972 Enamine

1-butanoylpiperidine-3-carboxylic acid

  • MDL number MFCD09813958
  • Cas Number : 1017213-97-6
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-51535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51535
Availability details for
Quantity
  • Availability
  • Add to Cart