ENA238712975 Enamine

1-(2-aminoethoxy)-2-methylbenzene hydrochloride

  • MDL number MFCD02667724
  • Cas Number : 313527-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51541

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51541
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart