ENA238712977 Enamine

4-(dimethyl-1,3-oxazol-2-yl)aniline

  • MDL number MFCD06739709
  • Cas Number : 708258-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-155
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51543
Availability details for
Quantity
  • Availability
  • Add to Cart