ENA238712979 Enamine

2-(2,3-dihydro-1H-isoindol-2-yl)acetic acid

  • MDL number MFCD04485858
  • Cas Number : 363165-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51545
Availability details for
Quantity
  • Availability
  • Add to Cart