ENA238712984 Enamine

5-ethylthiophene-2-sulfonyl chloride

  • MDL number MFCD06803451
  • Cas Number : 56921-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51551

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-51551
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart