ENA238712989 Enamine

1,1,3-trioxo-2-(prop-2-yn-1-yl)-2,3-dihydro-1lambda6,2-benzothiazole-6-carboxylic acid

  • MDL number MFCD12090921
  • Cas Number : 1156750-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51567
Availability details for
Quantity
  • Availability
  • Add to Cart