ENA238712992 Enamine

2-(phenylmethanesulfonamido)propanoic acid

  • MDL number MFCD01817160
  • Cas Number : 688789-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50336
Availability details for
Quantity
  • Availability
  • Add to Cart