ENA238712996 Enamine

3-[(1-methyl-1H-1,3-benzodiazol-2-yl)amino]propan-1-ol

  • MDL number MFCD05739208
  • Cas Number : 600180-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-51641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51641
Availability details for
Quantity
  • Availability
  • Add to Cart