ENA238712998 Enamine

2-[(1-ethyl-1H-1,3-benzodiazol-2-yl)amino]ethan-1-ol

  • MDL number MFCD04374076
  • Cas Number : 86978-98-5
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51643
Availability details for
Quantity
  • Availability
  • Add to Cart