ENA238712999 Enamine

1-benzyl-4-hydroxy-4-(4-methyl-1,3-thiazol-2-yl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD12912990
  • Cas Number : 1181521-21-0
  • Molecular Weight : 332.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51645
Availability details for
Quantity
  • Availability
  • Add to Cart