ENA238713000 Enamine

5-amino-N-(propan-2-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD11209997
  • Cas Number : 1187860-48-5
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-N-isopropyl-1H-pyrazole-4-carboxamide
Inchi Code 1S/C7H12N4O/c1-4(2)10-7(12)5-3-9-11-6(5)8/h3-4H,1-2H3,(H,10,12)(H3,8,9,11)
Inchi Key WVLRJCSZPLIHMA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-51646

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-51646
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart