ENA238713001 Enamine

methyl 2-(4-carbamoylpiperidin-1-yl)acetate

  • MDL number MFCD11176785
  • Cas Number : 1040020-69-6
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51648
Availability details for
Quantity
  • Availability
  • Add to Cart