ENA238713004 Enamine

2-(1H-pyrazol-1-yl)ethan-1-ol

  • MDL number MFCD00046076
  • Cas Number : 6314-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51651
Availability details for
Quantity
  • Availability
  • Add to Cart