ENA238713009 Enamine

N-(4-aminophenyl)-2-iodobenzamide

  • MDL number MFCD02271073
  • Cas Number : 926265-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51656
Availability details for
Quantity
  • Availability
  • Add to Cart