ENA238713012 Enamine

1-(5-methylthiophene-2-carbonyl)piperazine hydrochloride

  • MDL number MFCD12912995
  • Cas Number : 1193389-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51659
Availability details for
Quantity
  • Availability
  • Add to Cart