ENA238713013 Enamine

2-cyano-N-(prop-2-yn-1-yl)acetamide

  • MDL number MFCD09930675
  • Cas Number : 268222-23-7
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50338
Availability details for
Quantity
  • Availability
  • Add to Cart