ENA238713015 Enamine

N-(3-aminopropyl)-2-fluoro-N-methylaniline

  • MDL number MFCD10031317
  • Cas Number : 1018650-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51664
Availability details for
Quantity
  • Availability
  • Add to Cart