ENA238713018 Enamine

N-(3-amino-4-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxamide hydrochloride

  • MDL number MFCD12912840
  • Cas Number : 1193390-10-1
  • Molecular Weight : 288.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-50339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50339
Availability details for
Quantity
  • Availability
  • Add to Cart